Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD38 Mouse Monoclonal Antibody (Biotin) [clone: AT2]
Anti-CD38 Mouse Monoclonal Antibody (Biotin) [clone: AT2]
Supplier:  Biotium
Certificates
Anti-CD38 Mouse Monoclonal Antibody (Biotin) [clone: AT2]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB1086-500
  • BNCB1086-100
  • Antigen symbol:
  • CD38
  • CD38
  • Antigen synonyms:
  • cADP-ribose Hydrolase 1|ADP Ribosyl Cyclase 1|T10|NIM-R5 Antigen|CD38H|Acute Lymphoblastic Leukemia Cells Antigen|p45|NAD( ) Nucleosidase
  • cADP-ribose Hydrolase 1|ADP Ribosyl Cyclase 1|T10|NIM-R5 Antigen|CD38H|Acute Lymphoblastic Leukemia Cells Antigen|p45|NAD( ) Nucleosidase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone AT2
  • Clone AT2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 479214
  • 479214
  • Isotype:
  • IgG3 kappa
  • IgG3 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together