Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-VEGFA Mouse Monoclonal Antibody (CF405S) [clone: VEGF/1063]
 :  Biotium
 
Anti-VEGFA Mouse Monoclonal Antibody (CF405S) [clone: VEGF/1063]
 :  Biotium

 

  • Catalog No:
  • BNC041063-500
  • BNC041063-100
  • Antigen symbol:
  • VEGFA
  • VEGFA
  • Antigen name:
  • Vascular endothelial growth factor A
  • Vascular endothelial growth factor A
  • Antigen synonyms:
  • Vascular Endothelial Growth Factor A (VEGF-A)|MVCD1|VEGFA|Vascular Permeability Factor (VPF)
  • Vascular Endothelial Growth Factor A (VEGF-A)|MVCD1|VEGFA|Vascular Permeability Factor (VPF)
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone VEGF/1063
  • Clone VEGF/1063
  • Reactivity:
  • Rabbit, Human, Dog, Rat, Mouse
  • Rabbit, Human, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 73793
  • 73793
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together