Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC15A4 Rabbit Polyclonal Antibody
Anti-SLC15A4 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-59636
Supplier:  Novus Biologicals
Anti-SLC15A4 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-59636
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 15 Member A4
  • Antigen symbol:
    Solute carrier family 15, member 4
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    50 μg
  • Pk:
    50 µl

Specifications

About this item

Rabbit Polyclonal SLC15A4 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 4-8 ?g/ml

Type: Primary
Antigen: SLC15A4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human