Anti-DMC1 Mouse Monoclonal Antibody [clone: 1D12/4]
Mouse Monoclonal DMC1 Antibody (1D12/4). Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.
Type: Primary
Antigen: DMC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- NOVUNB100-181
- NOVUNB100-181SS
- Antigen symbol:
- DMC1
- DMC1
- Antigen name:
- DNA meiotic recombinase 1
- DNA meiotic recombinase 1
- Antigen synonyms:
- disrupted meiotic cDNA 1 homolog|HsLim15|DMC1H|dJ199H16.1|Meiotic recombination protein DMC1/LIM15 homolog|DMC1 homologue|DMC 1
- disrupted meiotic cDNA 1 homolog|HsLim15|DMC1H|dJ199H16.1|Meiotic recombination protein DMC1/LIM15 homolog|DMC1 homologue|DMC 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1D12/4
- 1D12/4
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 11144
- 11144
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together