Anti-WRN Rabbit Polyclonal Antibody
Catalog # NOVUNB100-471
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Werner syndrome, RecQ helicase-like
- Antigen symbol:WRN
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:RECQL3|RECQ3|RECQL2
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rabbit Polyclonal WRN Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: WRN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human