Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Double Stranded DNA Mouse Monoclonal Antibody [clone: AE-2]
Anti-Double Stranded DNA Mouse Monoclonal Antibody [clone: AE-2]
Catalog # BNUM0946-50
Supplier:  Biotium
Anti-Double Stranded DNA Mouse Monoclonal Antibody [clone: AE-2]
Catalog # BNUM0946-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Double Stranded DNA
  • Clonality:
    Monoclonal
  • Clone:
    Clone AE-2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG3 kappa
  • Reactivity:
    Human
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. This MAb recognizes the double stranded DNA in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in human cells. This MAb produces a homogeneous staining pattern in the nucleus of normal and malignant cells.,Double Stranded deoxyribonucleic acid (ds DNA) is the genetic material of all cells and many viruses and is a polymer of nucleotides. The monomer consists of phosphorylated 2-deoxyribose N-glycosidically linked to one of four bases, adenine, cytosine, guanine or thymine. These are linked together by 3',5'-phosphodiester bridges. In the Watson-Crick double-helix model, two complementary strands are wound in a right-handed helix and held together by hydrogen bonds between complementary base pairs.

  • Purified, BSA-free

Type: Primary
Antigen: Double Stranded DNA
Clonality: Monoclonal
Clone: AE-2
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human