Anti-RBP1 Mouse Monoclonal Antibody (CF640R) [clone: RBP/872]
: Biotium
Recognizes a protein of 21 kDa-25 kDa, identified as retinol binding protein-1 (RBP1). This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post-transnationally and results in defective delivery and supply to the epidermal cells.
Type: Primary
Antigen: RBP1
Clonality: Monoclonal
Clone: RBP/872
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Goat, Human, Monkey, Mouse, Rabbit, Rat
- Catalog No:
- BNC400872-500
- BNC400872-100
- Antigen symbol:
- RBP1
- RBP1
- Antigen name:
- Retinol binding protein 1
- Retinol binding protein 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone RBP/872
- Clone RBP/872
- Reactivity:
- Rabbit, Monkey, Goat, Human, Rat, Mouse
- Rabbit, Monkey, Goat, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 529571
- 529571
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Frequently Bought Together