Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CDKN1B Mouse Monoclonal Antibody (Biotin) [clone: DCS-72.F6]
Anti-CDKN1B Mouse Monoclonal Antibody (Biotin) [clone: DCS-72.F6]
Supplier:  Biotium
Certificates
Anti-CDKN1B Mouse Monoclonal Antibody (Biotin) [clone: DCS-72.F6]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0771-100
  • BNCB0771-500
  • Antigen symbol:
  • CDKN1B
  • CDKN1B
  • Antigen name:
  • Cyclin-dependent kinase inhibitor 1B
  • Cyclin-dependent kinase inhibitor 1B
  • Antigen synonyms:
  • CDKN1B|CDKN4|KIP1|Cyclin-dependent kinase inhibitor p27 Kip1|MEN1B|Cyclin Dependent Kinase Inhibitor 1B|MEN4
  • CDKN1B|CDKN4|KIP1|Cyclin-dependent kinase inhibitor p27 Kip1|MEN1B|Cyclin Dependent Kinase Inhibitor 1B|MEN4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone DCS-72.F6
  • Clone DCS-72.F6
  • Reactivity:
  • Monkey, Human, Rat, Mouse
  • Monkey, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 238990
  • 238990
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together