Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IPO-38 Mouse Monoclonal Antibody [clone: IPO-38]
 :  Biotium
 
Anti-IPO-38 Mouse Monoclonal Antibody [clone: IPO-38]
 :  Biotium

 

  • Catalog No:
  • BNUB0697-100
  • BNUB0697-500
  • Antigen name:
  • IPO-38
  • IPO-38
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone IPO-38
  • Clone IPO-38
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgM kappa
  • IgM kappa
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

 

Frequently Bought Together