Anti-IL17RA Rabbit Polyclonal Antibody
Catalog # BOSSBS-2606R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17 Receptor A
- Antigen symbol:IL17RA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:23765
- Antigen synonyms:IL-17 R|IL-17 RA|Il17r|AW538159|Cdw217|VDw217
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:240-280/866
- Purification:Purified by Protein A
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat