About this item
This MAb recognizes a single chain, transmembrane, heavily glycosylated protein of 90-120 kDa, which is identified as CD34. On the basis of differential sensitivity to degradation by specific enzymes, epitopes of monoclonal antibodies to CD34 are classified intothree main categories, class I, class II and class III. It is a class II antibody whose epitope is resistant to neuraminidase but sensitive to glycoprotease and chymopapain.CD34 expression is a hallmark for identifying pluripotent hematopoietic stem or progenitor cells. Its expression is gradually lost as lineage committed progenitors differentiate. CD34 is a marker of choice for staining blasts in acute myeloid leukemia. In addition, CD34 is expressed by soft tissue tumors, such as solitary fibrous tumor and gastrointestinal stromal tumor. Its expression is also found in vascular endothelium. Additionally, it appears that proliferating endothelial cells express this molecule more than the non-proliferating endothelial cells. Anti-CD34 labels > 85% of angiosarcoma and Kaposi's sarcoma, but with a lower specificity.
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEnd/10
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cynomolgus monkey, Human, Rhesus monkey
Specifications
- Catalog No:
- BNC940640-100
- BNC940640-500
- Antigen symbol:
- CD34
- CD34
- Antigen synonyms:
- Hematopoietic Progenitor Cell Antigen|Mucosialin|HPCA1
- Hematopoietic Progenitor Cell Antigen|Mucosialin|HPCA1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone QBEnd/10
- Clone QBEnd/10
- Reactivity:
- Cynomolgus monkey, Rhesus monkey, Human
- Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 374990
- 374990
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide