Fibroblast growth factor-1 (FGF-1), also designated acidic FGF, and fibroblast growth factor-2 (FGF-2), also designated basic FGF, are members of a family of growth factors that stimulate proliferation of cells of mesenchymal, epithelial and neuroectodermal origin. Additional members of the FGF family include the oncogenes FGF-3 (Int2) and FGF-4 (hst/Kaposi), FGF-5, FGF-6, FGF-7 (KGF), FGF-8 (AIGF), FGF-9 (GAF) and FGF-10 through FGF-23. Members of the FGF family share 30-55% amino acid sequence identity and similar gene structure, and are capable of transforming cultured cells when overexpressed in trans- fected cells. Cellular receptors for FGFs are members of a second multigene family, including four tyrosine kinases designated Flg (FGFR-1), Bek (FGFR-L), TKF and FGFR-3.
Type: Primary
Antigen: FGF23
Clonality: Monoclonal
Clone: FGF23/638
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
- Catalog No:
- BNC680638-500
- BNC680638-100
- Antigen symbol:
- FGF23
- FGF23
- Antigen name:
- Fibroblast growth factor 23
- Fibroblast growth factor 23
- Antigen synonyms:
- PHPTC|Phosphatonin|Tumor-derived hypophosphatemia-inducing factor|Fibroblast growth factor 23|ADHR|HPDR2|FGFN|FGF-23|HYPF
- PHPTC|Phosphatonin|Tumor-derived hypophosphatemia-inducing factor|Fibroblast growth factor 23|ADHR|HPDR2|FGFN|FGF-23|HYPF
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone FGF23/638
- Clone FGF23/638
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 287370
- 287370
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide