Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-DOCK2 Rabbit Polyclonal Antibody
  BOSSBS-12431R
 :  Bioss
Anti-DOCK2 Rabbit Polyclonal Antibody
  BOSSBS-12431R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    dedicator of cytokinesis 2
  • Antigen symbol:
    DOCK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    1794
  • Antigen synonyms:
    IMD40
  • Storage buffer:
    Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at −20 °C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4 °C
  • Immunogen:
    500-550/1830
  • Purification:
    Purified by Protein A
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Involved in cytoskeletal rearrangements required for lymphocyte migration in response of chemokines. Activates RAC1 and RAC2, but not CDC42, by functioning as a guanine nucleotide exchange factor (GEF), which exchanges bound GDP for free GTP. May also participate in IL2 transcriptional activation via the activation of RAC2.

Recommended Dilutions: IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: DOCK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat