Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MMP2 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Anti-MMP2 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
  BOSSBS-0412R-FITC
 :  Bioss
Anti-MMP2 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
  BOSSBS-0412R-FITC
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 2
  • Antigen symbol:
    MMP2
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    4313
  • Antigen synonyms:
    MMP-2|72 kDa gelatinase|matrix metalloproteinase-2|collagenase type IV-A|neutrophil gelatinase|gelatinase A|matrix metalloproteinase-II|72 kDa type IV collagenase
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4 °C for 12 months
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    20-70/476
  • Purification:
    Purified by Protein A
  • Size:
    100 μL
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat