Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MMP9 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Anti-MMP9 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # BOSSBS-0397R-FITC
Dodavatel:  Bioss
Anti-MMP9 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # BOSSBS-0397R-FITC
Dodavatel:  Bioss

Specifikace

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 9
  • Antigen symbol:
    MMP9
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    4318
  • Antigen synonyms:
    92kDa type IV collagenase)|MMP-9|GELB|92 kDa gelatinase|MANDP2|matrix metalloproteinase 9 (gelatinase B|92 kDa type IV collagenase|type V collagenase|matrix metalloproteinase-9|92kDa gelatinase|macrophage gelatinase|CLG4B
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4 °C for 12 months
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    630-680/707
  • Purification:
    Purified by Protein A
  • Size:
    100 μL
  • Pk:
    100 µl

Specifikace

O tomto produktu

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat