Reacts with a protein of 57 kDa, identified as the L1 protein of human papilloma virus type 16 (HPV-16). It is the major capsid protein of HPV-16. Infection with specific types of HPV has been associated with an increased risk of developing cervical neoplasia. HPV types 6 and 11 have been associated with relatively benign diseases such as genital warts but types 16 and 18 are strongly associated with cervical, vaginal, and vulvar malignancies. The antibody reacts very strongly with formalin-fixed, paraffin-embedded tissues containing HPV-16 or -33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV-6 or HPV-11. It cross-reacts with HPV37.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: CAMVIR-1
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: HPV
- Catalog No:
- BNC400502-100
- BNC400502-500
- Antigen name:
- Human Papillomavirus 16
- Human Papillomavirus 16
- Antigen synonyms:
- HPV-16 capsid|HPV16 L1|HPV-16|HPV16 major capsid protein L1|Human papillomavirus type 16 major capsid protein L1|Human papillomavirus type 16 L1
- HPV-16 capsid|HPV16 L1|HPV-16|HPV16 major capsid protein L1|Human papillomavirus type 16 major capsid protein L1|Human papillomavirus type 16 L1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone CAMVIR-1
- Clone CAMVIR-1
- Reactivity:
- Human papilloma virus (HPV)
- Human papilloma virus (HPV)
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide