Anti-CD5 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: Cris-1]
Catalog # BNUM0176-50
Supplier: Biotium
Specifications
- Antibody type:Primary
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:Clone Cris-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Rhesus monkey,Human
- Environmentally Preferable:
- Gene ID:58685
- Antigen synonyms:CD5 antigen (p56 62)|Ly12|LEU1|Lymphocyte glycoprotein T1/Leu1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|LyA
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Recognizes a 67 kDa transmembrane protein, which is identified as CD5 (HLDA I; WS Code T 29HLDA III; WS Code T 530). The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. Anti-CD5 is a pan T-cell marker that also reacts with a range of neoplastic B-cells, e.g. chronic lymphocytic leukemia/small lymphocytic lymphoma (CLL/SLL), mantle cell lymphoma, and a subset (~10%) of diffuse large B-cell lymphoma. CD5 aberrant expression is useful in making a diagnosis of mature T-cell neoplasms.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: Cris-1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rhesus monkey