Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-HIF1A Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HIF1A-84]
Catalog # BNUM0084-50
Supplier:  Biotium
Anti-HIF1A Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HIF1A-84]
Catalog # BNUM0084-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HIF1-alpha
  • Antigen symbol:
    HIF1A
  • Clonality:
    Monoclonal
  • Clone:
    Clone HIF1A-84
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Gene ID:
    597216
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

HIF1 (hypoxia-inducible factor 1), a heterodimeric transcription factor complex central to cellular response to hypoxia, consists of two subunits (HIF-1 alpha and HIF-1 beta) which are basic helix-loop-helix proteins of the PAS (Per, ARNT, Sim) family. Expression of HIF-1 alpha protein is regulated by cellular oxygen level alterations as well as in oxygen-independent manner via different cytokines (through the PI3K-AKT-mTOR pathway), growth factors, oncogenic activation, or loss of tumor suppressor function etc. In normoxic cells, HIF-1 alpha is proline hydroxylated leading to a conformational change that promotes its binding to the VLH (von Hippel Lindau) protein E3 ligase complex; ubiquitination and followed by rapid proteasomal degradation. Hypoxia as well as chemical hydroxylase inhibitors (desferrioxamine, cobalt etc.) inhibit HIF-1 alpha degradation and lead to its accumulation in the cells, whereas, contrastingly, HIF-1 beta/ARNT (AhR nuclear translocator) remains stable under both conditions. Besides their critical role in hypoxic response, HIF1s regulates the transcription of genes responsible for angiogenesis, erythropoiesis/iron-metabolism, glucose metabolism, cell proliferation/survival, adipogenesis, carotid body formation, B lymphocyte development and immune reactions.

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone: HIF1A-84
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human