Recognizes a 67 kDa glycoprotein, which is identified as CD33. It is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies; these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. CD33 is expressed on myeloid progenitors, monocytes, granulocytes, dendritic cells and mast cells. It is absent on platelets, lymphocytes, erythrocytes and hematopoietic stem cells.
Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: C33/68
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
- Catalog No:
- BNCB0068-500
- BNCB0068-100
- Antigen symbol:
- CD33
- CD33
- Antigen synonyms:
- gp67|Sialic acid-binding Ig-like lectin 3 (SIGLEC3)|My9|Myeloid cell surface antigen CD33|p67
- gp67|Sialic acid-binding Ig-like lectin 3 (SIGLEC3)|My9|Myeloid cell surface antigen CD33|p67
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone C33/68
- Clone C33/68
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 83731
- 83731
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide