This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It was reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18.
Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: C-04
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cat, Cow, Dog, Ferret, Goat, Hamster, Horse, Human, Marmoset, Monkey, Mouse, Pig, Rat, Sheep
- Catalog No:
- BNC880041-100
- BNC880041-500
- Antigen symbol:
- KRT18
- KRT18
- Antigen name:
- Cytokeratin 18
- Cytokeratin 18
- Antigen synonyms:
- K18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 Protein
- K18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 Protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone C-04
- Clone C-04
- Reactivity:
- Rat, Mouse, Cat, Marmoset, Hamster, Pig, Horse, Monkey, Sheep, Goat, Ferret, Human, Bovine, Dog
- Rat, Mouse, Cat, Marmoset, Hamster, Pig, Horse, Monkey, Sheep, Goat, Ferret, Human, Bovine, Dog
- Host:
- Mouse
- Mouse
- Gene ID:
- 406013
- 406013
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide