Order Entry
Czech Republic
ContactUsLinkComponent
DNA extraction kit, forensic, MPure™ 12, MP Biomedicals
DNA extraction kit, forensic, MPure™ 12, MP Biomedicals
Catalog # ICNA117022110
Supplier:  MP Biomedicals
CAS Number:  
undefined
DNA extraction kit, forensic, MPure™ 12, MP Biomedicals
Catalog # ICNA117022110
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Forensic samples
  • Target molecule:
    DNA
  • Description:
    MPure™ Forensic DNA Extraction Kit, 48 tests
  • Description:
    MPure™ Forensic DNA Extraction Kit, 48 tests

Specifications

About this item

MPure™ Forensic DNA Extraction Kit is used with the MPure-12™ aNAP system for the extraction and purification of genomic DNA from forensic samples.