Standardní roztok hliníku pro ICP-MS, ARISTAR®
Supplier: VWR Chemicals
Certificates
About this item
- vyrobeno z vysoce čistých kyselin, vody podle ASTM I 18 Megaohm
- roztok stanoven titrací
- konečná koncentrace ověřena podle referenčního materiálu NIST
- celková maximální nejistota měření: 0,5%
- dodáno s úplným certifikátem o analýze
1000 ppm ekvivalentní 1000 mg/l
Certifications: Certifikace SRM dle NIST Vyrobeno a testováno v zařízeních akreditovaných dle ISO 34/ ISO17025
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together