TwiXS packs, Acura® manual XS 826
Supplier: Socorex Isba
Certificates
About this item
These ready-to-use kits include two Acura® manual XS pipettes, one shelf pipette holder, Qualitips® samples, QC certificates and operating instructions.
Frequently Bought Together