Order Entry
Czech Republic
ContactUsLinkComponent
Bovine serum albumin (BSA)
Bovine serum albumin (BSA)
Supplier:  SEQENS
Certificates
undefined
Bovine serum albumin (BSA)
Supplier:  SEQENS

Specifications

  • Source:
    Serum
  • Species:
    Bovine
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Formulation:
    lyophilized
  • pH:
    5.2

Specifications

Related Information

Frequently Bought Together