ContactUsLinkComponent
Resistor kit for conductivity meters, Orion™
Resistor kit for conductivity meters, Orion™
Catalog # ORIO1010001
Supplier:  Thermo Orion
Resistor kit for conductivity meters, Orion™
Catalog # ORIO1010001
Supplier:  Thermo Orion
Specifications
  • Description:
    Conductivity calibration resistor kit with 8-pin MiniDIN connection, for use with Orion Star and Orion Star A Series conductivity meters
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This resistor calibration kit can be used to verify the performance of Thermo Scientific™ Orion™ Versa Star conductivity meters, Thermo Scientific™ Orion™ Star A series conductivity meters and legacy Orion™ Star series conductivity meters.
  • Conductivity calibration resistor kit with 8 pin MiniDIN connection
Delivery: Kit includes six resistors that simulate nominal conductance values between 1 µS and 100 mS.