ContactUsLinkComponent
Safety air outlet filter
Safety air outlet filter
Catalog # JOURJR-S-20004
Supplier:  VICI JOUR RESEARCH
Safety air outlet filter
Catalog # JOURJR-S-20004
Supplier:  VICI JOUR RESEARCH
Specifications
  • Description:
    Safety air outlet filter
  • Material:
    PP
  • Seq:
    06
  • Connector:
    1/4-28 valve to cap and female Luer filter connections
  • Pk:
    1
Specifications
About this item
This safety air outlet filter regulates the pressure during helium sparging.