Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Bcl-2 Mouse Monoclonal Antibody [clone: 83-8B]
Anti-Bcl-2 Mouse Monoclonal Antibody [clone: 83-8B]
Catalog # ENZOADIAAM072E
Supplier:  Enzo Life Sciences
Anti-Bcl-2 Mouse Monoclonal Antibody [clone: 83-8B]
Catalog # ENZOADIAAM072E
Supplier:  Enzo Life Sciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B-cell lymphoma 2
  • Antigen symbol:
    BCL2
  • Clonality:
    Monoclonal
  • Clone:
    83-8B
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Drosophila,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid
  • Antigen synonyms:
    PPP1R50|Bcl-2
  • Storage buffer:
    PBS, pH 7,2, containing 50% glycerol.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant rat Bcl-2 beta
  • Purification:
    Purified by Protein A affinity chromatography
  • Pk:
    100 µG

Specifications

About this item

The Bcl-2 family of proteins regulate the intrinsic apoptotic pathway and are characterized by the sharing of at least one of four conserved Bcl-2 Homology (BH) domains.

  • Dilution Range: 10 µg/ml for Flow Cytometry, 1 µg/ml, ECL for Western blot
  • Detects a band of ~26kDa by Western blot

Bcl-2 family members function as either pro- or anti-apoptotic factors, and are generally categorized into three groups based on structural homology and function. Bcl-2-like proteins (Bcl-2, Bcl-xL, Bcl-w, A1, and Mcl-1) are pro-survival. Pro-apoptotic members can be sub-divided into two groups: the multi-BH domain group, including Bax, Bak, and Bok; and the more numerous and diverse BH3-only group, which includes Bim, Bad, and Bid.

Type: Primary
Antigen: B-cell lymphoma 2
Clonality: Monoclonal
Clone: 83-8B
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Drosophila, Human, Mouse, Rat