Sample loops for high pressure valves, stainless steel, Upchurch Scientific®
UPCH1876
- Kapacita:10000 µl
- Popis:Sample loop
- Materiál:Nerezová ocel
- Sekvence:01
- Bal.:1
- Pro zkumavky s vnitř. Ø:2,0 mm
- Pro zkumavky s vnějš. Ø:3,1 mm
These high quality stainless steel sample loops have burr-free, square-cut ends to ensure a flush connection to valve ports.
- Size designations of loops are nominal
- Supplied with fittings that are not swaged onto the tube
The loop has to be completely bottomed in the injector port before the ferrule is swaged onto the tube.