ContactUsLinkComponent
Hubbel, Mendel and Wakeman Salt Mixture, MP Biomedicals
Hubbel, Mendel and Wakeman Salt Mixture, MP Biomedicals
Catalog # ICNA0290283802
Dodavatel:  MP Biomedicals
Hubbel, Mendel and Wakeman Salt Mixture, MP Biomedicals
Catalog # ICNA0290283802
Dodavatel:  MP Biomedicals
Specifikace
  • Description:
    Hubbel, Mendel and Wakeman salt mixture
  • Pk:
    2 kg
Specifikace
O tomto produktu
Hubbel, Mendel and Wakeman powder does not contain zinc, is low in manganese and phosphorus, and has a high calcium to phosphorus ratio.
Designed for use in formulating animal research diets, especially with relatively high casein levels.
For complete composition see product datasheet.