


Xylene (mixture of isomers), clear, colourless liquid ACS
Catalog # ICNA0215869390
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Assay:≥98.5% plus ethylbenzene (not to exceed 25%)
- Color (APHA):≤10
- Residue After Evaporation:≤0.002%
- Substances Darkened by Sulfuric Acid :Passes Test
- Sulfur Compounds:≤0.003%
- Water:≤0.05%
Specification Test Results
About this item
:
:106.17 g/mol
Boiling Pt:137…140 °C (lit.)
Melting Pt:–47.9…+13.3 °C (lit.)
Density:0.86 g/mL (25 °C) (lit.)
Storage Temperature:Ambient
UN:1307
ADR:3,III
:GHS02
:GHS07
:GHS08
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264