Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
1-Hexanol, liquid

1-Hexanol, liquid

100 mL
Catalog # ICNA0215736280
Supplier:   MP Biomedicals
1-Hexanol, liquid
Catalog # ICNA0215736280
Supplier:  MP Biomedicals
CAS Number:  111-27-3
1-Hexanol, liquid
1-Hexanol, liquid

100 mL

CAS Number:  111-27-3
Price
581,00 Kč
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
  • CAS Number:111-27-3
  • Formula:H₃C(CH₂)₄CH₂OH
  • MDL Number:MFCD00002982
  • Molecular Weight:102.18 g/mol
  • UN:2282
  • ADR:3,III
Specifications

Specifications

  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.