2,2-Diphenyl-1-picrylhydrazyl (free radical) ≥95%, crystalline solid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:1898-66-4
:
Formula:C₁₈H₁₂N₅O₆
MDL Number:MFCD00007231
:394.32 g/mol
EINECS:217-591-8
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Purity:≥95%
- Stability:≥2 years at −20 °C
- Form:Crystalline Solid
- UV/Vis:λmax: 327, 515 nm
Specification Test Results
Frequently Bought Together