


Cetrimonium bromide, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:57-09-0
:
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772
:364.45 g/mol
Storage Temperature:Ambient
UN:3077
ADR:9,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Identity Test Passes:Passes
- FTIR:Conforms to Standard
- Storage:Store at Room Temperature (15 to 30 °C)
- Aggregation Number:170
- CMC:1 mM (20 - 25 °C)
- pH:6,0 - 7,5 (0,1 M H2O, 0 °C) (Lit.)
Specification Test Results
Frequently Bought Together