


Benzalkoniumchlorid ~97%, waxy solid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:63449-41-2
:8001-54-5
Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
MDL Number:MFCD00145757
Storage Temperature:Ambient
UN:2923
ADR:8,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Identity Test:Passes
- Purity:~97%
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together