
Lithium chloride ≥99%, white powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
:42.39 g/mol
Melting Pt:605 °C (lit.)
:GHS07
Specification Test Results
- Assay:≥99%
- Insoluble Matter:≤0,01%
- Titrable Base:≤0,008 mEq/g
- Loss on Drying:≤1,0%
- Nitrate (NO₃):≤0,001%
- Sulphate (SO₄):≤0,01%
- Barium (Ba):≤0,003%
- Heavy Metals:≤0,002%
- Iron (Fe):<0,001%
- Calcium (Ca):≤0,01%
- Potassium (K):≤0,01%
- Sodium (Na):≤0,20%
Specification Test Results
Frequently Bought Together