Lithium carbonate, powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:554-13-2
:
Formula:Li₂CO₃
MDL Number:MFCD00011084
:73.89 g/mol
Specification Test Results
- Synonyms:Carbonic Acid Dilithium Salt; Carbonic Acid Lithium Salt; Dilthium Carbonate; Lithiumcarbonat; Carbolithium
- Form:White Powder
- Storage:Store at Room Temperature (15 to 30 °C)
- Assay:≥99%
- Heavy Metals:≤0,002% (as Pb)
- Insoluble in Dilute HCl:≤0,01%
- Chloride:≤0,005%
- Nitrate (NO3):≤5 ppm
- Sulphur Compounds (as SO4):≤0,2%
- Iron (Fe):≤0,002%
- Calcium (Ca):≤0,01%
- Potassium (K):≤0,01%
- Sodium (Na):≤0,1%
Specification Test Results
Frequently Bought Together