
Sodium diphenylamine-4-sulphonate, powder ACS redox indicator
Catalog # ICNA0215251905
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Sensitivity as Indicator:Passes Test
Specification Test Results
About this item
:
:271.27 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:6152-67-6
Formula:C₁₂H₁₀NNaO₃S
MDL Number:MFCD00036468