Order Entry
Czech Republic
ContactUsLinkComponent
HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid) ≥99%, white powder
HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid) ≥99%, white powder
Supplier:  MP Biomedicals
Certificates
HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid) ≥99%, white powder
Supplier:  MP Biomedicals
Certificates

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • pH (1% aq soln) :
    Record Result

Specification Test Results

Related Information

Frequently Bought Together