Order Entry
Czech Republic
ContactUsLinkComponent
Lithium dodecyl sulphate ≥99%, white powder, Ultrapure
Flammable
Corrosive
Irritant
Lithium dodecyl sulphate ≥99%, white powder, Ultrapure
Supplier:  MP Biomedicals
Certificates
Flammable
Corrosive
Irritant
Lithium dodecyl sulphate ≥99%, white powder, Ultrapure
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • OD260 (1% aq soln):
    ≤0.1
  • OD280 (1% aq soln):
    ≤0.1

Specification Test Results

Frequently Bought Together