ContactUsLinkComponent
Methyl red, dark red powder ACS
Methyl red, dark red powder ACS
Supplier:  MP Biomedicals
Certificates
Methyl red, dark red powder ACS
Supplier:  MP Biomedicals
Certificates
Specification Test Results
  • Melting Point:
    179 - 182 °C
  • Clarity of Alcohol Solution:
    Passes Test
  • Visual Transition Interval:
    From pH 4.2 (pink) to 6.2 (yellow)