Order Entry
Czech Republic
ContactUsLinkComponent
 
Corn Syrup, Liquid, MP Biomedicals
Catalog # ICNA0210141301
Supplier:  MP Biomedicals
Corn Syrup, Liquid, MP Biomedicals
Catalog # ICNA0210141301
Supplier:  MP Biomedicals

Specifications

  • Description:
    Corn syrup, liquid
  • Pk:
    3,79 L

Specifications

About this item

Corn-syrup is a dual acid-enzyme conversion syrup which contains approximately three times as much maltose as regular corn syrups processed to the same degree of conversion using acid only.

  • Conversion: acid-enzyme
  • Maltose Content: ~30%

Corn syrup is used in commercially prepared foods as a thickener, a sweetener and as a humectant - an ingredient that retains moisture and thus maintains a food's freshness.

Caution: Research Use Only (RUO).