
3-(4-Bromophenyl)propan-1-ol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25574-11-2
:
Formula:C₉H₁₁BrO
MDL Number:MFCD09028724
:215.09 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (GC):97,5%
Specification Test Results
Frequently Bought Together