Methyl palmitate ≥99%, white crystals
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:112-39-0
:
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00008994
:270.46 g/mol
Melting Pt:32…34 °C (lit.)
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
Specification Test Results
Frequently Bought Together