Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Mouse ANGPTL3 ELISA kit
Mouse ANGPTL3 ELISA kit
Mouse ANGPTL3 ELISA kit
Catalog # BIRBORB79219-96
Supplier:
Biorbyt
CAS Number:
Expand image
Mouse ANGPTL3 ELISA kit
Catalog # BIRBORB79219-96
Supplier:
Biorbyt
CAS Number:
Specifications
Pk:
96
Primary antibody reactivity:
Mouse
Description:
Mouse ANGPTL3 ELISA Kit
Specifications
About this item
Product Details
Documentation
Product Details
Documentation