ContactUsLinkComponent
Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, yellow crystalline powder ACS
Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, yellow crystalline powder ACS
Supplier:  MP Biomedicals
Certificates
Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, yellow crystalline powder ACS
Supplier:  MP Biomedicals
Specification Test Results
  • Assay:
    98,5 - 102,0%
  • Insoluble Matter:
    ≤0,005%
  • Chloride:
    ≤0,01%
  • Sulphate :
    Passes Test