di-Potassium hydrogen phosphate, anhydrous ≥98%, white crystalline powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7758-11-4
:
Formula:K₂HPO₄
MDL Number:MFCD00011383
:174.18 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay:≥98%
- pH (5% aq soln):8,5 - 9,6
- Insoluble Matter:≤0,01%
- Loss on Drying:≤1,0%
- Chloride (Cl):≤0,003%
- Nitrogen (as N):≤0,001%
- Sulfate (SO₄):≤0,005%
- Heavy Metals (as Pb):≤5 ppm
- Iron (Fe):≤0,001%
- Sodium (Na):≤0,05%
Specification Test Results
Frequently Bought Together