Guanidinium hydrochloride, white powder, Practical grade
:50-01-1
:
:NH₂C(=NH)NH₂·HCl
:MFCD00013026
:95.53 g/mol
- Identity Test:Passes
- Melting Point:175 - 190 °C
- FTIR:Conforms to Standard
- Loss on Drying:≤1,0%
- Solubility (6M aq soln) R:Relatively Clear to Cloudy Yellow Solution with Sediment on Standing
Frequently Bought Together