ContactUsLinkComponent
Vector, pBR322, MP Biomedicals
Vector, pBR322, MP Biomedicals
Catalog # ICNA04821225
Supplier:  MP Biomedicals
CAS Number:  
Vector, pBR322, MP Biomedicals
Catalog # ICNA04821225
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    25 µG
  • Description:
    Vector, pBR322
Specifications
About this item
pBR322 is a 4363 bp vector purified from Escherichia coli. It is one of the most commonly used cloning vectors, that confers resistance to ampicillin and tetracycline. It contains single restriction sites for Pst I and Pvu I within the ampicillin-resistance gene (ampr) and single restriction sites for BamH I, Sal I, Sph I, EcoR V, Nru I and Xma III within the tetracycline-resistance gene (tetr).
  • 4363 bp vector Purified from Escherichia coli Contains single restriction sites
Plasmid pBR322 was one of the first multipurpose cloning vectors constructed for use in Escherichia coli. This plasmid and derivatives have been used for a number of purposes including cloning, selection and expression of recombinant molecules, construction of shuttle vectors and vectors for nucleotide sequencing, studies of elements involved in gene expression, as plasmid DNA standards, and as a model system for studies on prokaryotic plasmid replication.
Note: Elevated temperatures and/or multiple freeze/thaw cycles will cause some of the ccDNA to convert to ocDNA.