ContactUsLinkComponent
Accessories for S-Monovette®, ESR (Sodium citrate)
Accessories for S-Monovette®, ESR (Sodium citrate)
Catalog # SARS90.1090
Supplier:  SARSTEDT
Accessories for S-Monovette®, ESR (Sodium citrate)
Catalog # SARS90.1090
Supplier:  SARSTEDT
Specifications
  • Description:
    Sedimentation rack with gradutated back wall for S-Sedivette®
  • Pk:
    1