Disposable syringes
Supplier: THERMO NATIONAL SCIENTIFIC
Jump to:
About this item
These syringes are an ideal choice for all syringe filter applications. They feature a two-part construction using PP barrels and PE plungers. This eliminates the need for rubber or synthetic plunger gaskets and requires no silicone or oil lubricant in the barrel.
- Non sterile
- Bulk packed
4 Options Available Below
Product Details & Documents
These syringes are an ideal choice for all syringe filter applications. They feature a two-part construction using PP barrels and PE plungers. This eliminates the need for rubber or synthetic plunger gaskets and requires no silicone or oil lubricant in the barrel.
- Non sterile
- Bulk packed
Product Family Options
Product Information
- VolumeAvailabilityPrice
- NSCAS7515-3Luer lock syringe
- NSCAS7515-10Luer lock syringe
- NSCAS7515-20Luer lock syringe
- NSCAS7510-1Luer slip syringe